siRNA Details
Gene Symbol: MAGEA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005365.4 | NP_005356.1 | 4 | 603 | ||
XM_005262334.3 | XP_005262391.1 | 4 | 625 | |||
XM_005262335.3 | XP_005262392.1 | 3 | 532 | |||
GenBank mRNA | AK292272.1 | - | 4 | 610 | ||
AK313321.1 | - | 4 | 610 | |||
AY310325.1 | - | 1 | 323 | |||
BC002351.2 | - | 4 | 603 | 2820966 | ||
BC098100.1 | - | 4 | 564 |
Gene Symbol: MAGEA9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080790.1 | NP_001074259.1 | 4 | 610 | ||
XM_005278192.3 | XP_005278249.1 | 4 | 625 | |||
XM_005278193.3 | XP_005278250.1 | 3 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
