siRNA Details
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 2 | 299 | ||
NM_001313893.1 | NP_001300822.1 | 2 | 1059 | |||
NM_002128.5 | NP_002119.1 | 2 | 302 | |||
GenBank mRNA | AA430980.1 | - | 2 | 303 | ||
AB208998.1 | - | 2 | 186 | |||
AK057120.1 | - | 2 | 236 | |||
AK122825.1 | - | 2 | 1061 | |||
AK291494.1 | - | 2 | 273 | |||
AK304506.1 | - | 2 | 273 | |||
AK310975.1 | - | 2 | 248 | |||
AV763680.1 | - | 2 | 299 | |||
AX795278.1 | - | 2 | 194 | |||
AY377859.1 | - | 1 | 118 | |||
BC003378.1 | - | 2 | 194 | 2901382 | ||
BC030981.1 | - | 2 | 213 | 4041682 | ||
BC066889.1 | - | 2 | 197 | 5268698 | ||
BC067732.1 | - | 2 | 217 | 4818524 | ||
BC141844.1 | - | 2 | 195 | 8860319 | ||
BT006940.1 | - | 1 | 118 | |||
BT020159.1 | - | 1 | 118 | |||
BX647267.1 | - | 1 | 682 | |||
CB051582.1 | - | 2 | 310 | |||
CR456863.1 | - | 1 | 118 | |||
CR749614.1 | - | 2 | 290 | |||
D63874.1 | - | 2 | 194 | |||
X12597.1 | - | 2 | 170 |
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 153 | 9021482 | |
BC137483.1 | - | 1 | 153 | 9021483 | ||
DQ778130.1 | - | 4 | 658 | |||
DQ778131.1 | - | 3 | 518 |
Gene Symbol: HMGB1P34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC068102.1 | - | 1 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
