siRNA Details
Gene Symbol: BANF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014977.3 | NP_001014977.2 | 2 | 162 | ||
NM_001159495.1 | NP_001152967.1 | 2 | 86 | |||
NM_178477.4 | NP_848572.3 | 3 | 325 | |||
XM_005260668.3 | XP_005260725.1 | 4 | 698 | |||
XM_011529170.1 | XP_011527472.1 | 3 | 535 | |||
XM_011529171.2 | XP_011527473.1 | 3 | 337 | |||
GenBank mRNA | AA394107.1 | - | 2 | 125 | ||
AW022487.1 | - | 2 | 94 | |||
BC054871.1 | - | 2 | 166 | 6592641 | ||
BC062735.1 | - | 3 | 316 | 6591888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
