siRNA Details
Gene Symbol: MT1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC103840.1 | - | 1 | 178 | ||
BC133653.1 | - | 1 | 178 |
Gene Symbol: MT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001447.2 | 3 | 226 | |||
GenBank mRNA | BC070351.1 | - | 3 | 226 | ||
BG747999.1 | - | 3 | 200 | |||
X97261.1 | - | 3 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
