siRNA Details
Gene Symbol: SLC35E2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110781.2 | NP_001104251.1 | 2 | 620 | ||
NM_001290264.1 | NP_001277193.1 | 3 | 1038 | |||
XM_011542072.1 | XP_011540374.1 | 1 | 229 | |||
GenBank mRNA | AB007916.3 | - | 2 | 558 | ||
AK125619.1 | - | 2 | 544 | |||
AK307886.1 | - | 1 | 222 | |||
BC110653.1 | - | 2 | 596 | 4392812 | ||
CR936618.1 | - | 3 | 758 |
Gene Symbol: SLC35E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199787.1 | NP_001186716.1 | 3 | 958 | ||
NM_182838.2 | NP_878258.1 | 2 | 540 | |||
GenBank mRNA | AL049665.1 | - | 2 | 533 | ||
BC101666.1 | - | 1 | 139 | 8069172 | ||
BC112289.1 | - | 1 | 139 | 8327757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
