siRNA Details
Gene Symbol: SLURP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020427.2 | NP_065160.1 | 1, 2 | 81 | ||
GenBank mRNA | AY579080.1 | - | 1, 2 | 81 | ||
BC069292.1 | - | 1, 2 | 81 | 7262730 | ||
BC105133.1 | - | 1, 2 | 78 | 8144136 | ||
BC105135.1 | - | 1, 2 | 76 | 8144138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
