siRNA Details
Gene Symbol: CTAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020994.4 | NP_066274.2 | 2 | 395 | ||
NM_172377.4 | NP_758965.2 | 2 | 395 | |||
GenBank mRNA | AJ012833.1 | - | 2 | 300 | ||
AJ012834.1 | - | 2 | 384 | |||
AJ012835.1 | - | 2 | 386 | |||
AJ223040.1 | - | 2 | 395 | |||
AJ223041.1 | - | 2 | 383 | |||
AY679088.1 | - | 2 | 383 | |||
AY679089.1 | - | 2 | 383 | |||
BC002833.2 | - | 2 | 349 | 3638129 | ||
BC113998.1 | - | 2 | 375 | 40081511 | ||
BC114933.1 | - | 3 | 158 | |||
BC128045.1 | - | 2 | 375 | 40081514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
