siRNA Details
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033257.3 | NP_150282.2 | 2 | 260 | ||
GenBank mRNA | AF228708.1 | - | 2 | 240 | ||
AK001528.1 | - | 2 | 201 | |||
AK289952.1 | - | 2 | 178 | |||
AK307210.1 | - | 2 | 196 | |||
AK310899.1 | - | 2 | 235 | |||
BC000682.2 | - | 2 | 196 | 3349309 | ||
CR456361.1 | - | 2 | 165 | |||
KJ535014.1 | - | 2 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
