siRNA Details
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 5 | 848 | ||
XM_011530394.1 | XP_011528696.1 | 6 | 835 | |||
GenBank mRNA | AF228707.1 | - | 5 | 792 | ||
AK098780.1 | - | 5 | 1070 | |||
BC047039.1 | - | 5 | 741 | 5229172 | ||
CR456434.1 | - | 5 | 718 | |||
X96484.1 | - | 5 | 710 |
Gene Symbol: DGCR6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033257.3 | NP_150282.2 | 5 | 813 | ||
GenBank mRNA | AF228708.1 | - | 5 | 793 | ||
AK001528.1 | - | 5 | 754 | |||
AK289952.1 | - | 5 | 731 | |||
AK307210.1 | - | 4 | 648 | |||
AK310899.1 | - | 4 | 1299 | |||
BC000682.2 | - | 5 | 749 | 3349309 | ||
CR456361.1 | - | 5 | 718 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 5 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
