siRNA Details
Gene Symbol: VCX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001888.3 | NP_001001888.3 | 3 | 1114 | ||
GenBank mRNA | AF167080.2 | - | 4 | 1024 | ||
BC098143.1 | - | 4 | 736 | 40014420 |
Gene Symbol: VCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013452.2 | NP_038480.2 | 3 | 906 | ||
XM_011545490.2 | XP_011543792.1 | 3 | 738 | |||
GenBank mRNA | AF159129.1 | - | 2 | 713 | ||
AF167081.2 | - | 3 | 906 | |||
BC098123.1 | - | 4 | 712 | 40002642 | ||
BC098149.1 | - | 3 | 592 | 40002644 | ||
BC126903.1 | - | 2 | 803 | 40030361 |
Gene Symbol: VCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016378.3 | NP_057462.2 | 3 | 754 | ||
GenBank mRNA | AF159127.1 | - | 2 | 472 | ||
AF167079.1 | - | 3 | 754 | |||
BC096715.1 | - | 2 | 472 | 40002645 | ||
BC098101.1 | - | 2 | 472 | 40002641 | ||
BC098113.1 | - | 2 | 472 | 40014419 | ||
BC098163.1 | - | 2 | 472 | 40014421 | ||
BC104153.2 | - | 2 | 472 | 40029400 | ||
BC104167.1 | - | 2 | 563 | 40030354 | ||
BC104168.1 | - | 2 | 563 | 40030355 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016379.3 | NP_057463.2 | 3 | 934 | ||
XM_006724494.3 | XP_006724557.1 | 3 | 658 | |||
GenBank mRNA | AF159128.1 | - | 2 | 633 | ||
AF167078.1 | - | 3 | 934 | |||
BC039722.1 | - | 2 | 645 | |||
BC104154.2 | - | 2 | 441 | 40029402 | ||
BC126902.1 | - | 4 | 743 | 40030357 |
Gene Symbol: VCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004679.2 | NP_004670.1 | 2 | 488 | ||
GenBank mRNA | AF000979.1 | - | 2 | 488 | ||
BC096729.1 | - | 2 | 441 | 40014422 |
Gene Symbol: VCY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181880.1 | NP_870996.1 | 2 | 484 | ||
GenBank mRNA | BC056508.1 | - | 2 | 484 | 6450744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
