siRNA Details
Gene Symbol: RBMY1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303410.1 | NP_001290339.1 | 4 | 346 | ||
NM_152585.2 | NP_689798.1 | 4 | 346 | |||
XM_017030033.1 | XP_016885522.1 | 3 | 162 | |||
GenBank mRNA | BC030018.2 | - | 4 | 346 | 5269012 | |
X76060.1 | - | 4 | 328 |
Gene Symbol: RBMY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320944.1 | NP_001307873.1 | 4 | 386 | ||
NM_005058.3 | NP_005049.1 | 4 | 386 | |||
XM_011531437.2 | XP_011529739.1 | 4 | 325 | |||
XM_011531438.1 | XP_011529740.1 | 4 | 325 | |||
XM_017030060.1 | XP_016885549.1 | 3 | 162 | |||
GenBank mRNA | BC070298.1 | - | 4 | 351 | 6615329 | |
BC146853.1 | - | 4 | 299 | 9056781 | ||
BU568309.1 | - | 4 | 354 | |||
X76059.1 | - | 4 | 351 |
Gene Symbol: RBMY1A3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001547.1 | 3 | 239 |
Gene Symbol: RBMY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006121.3 | NP_001006121.1 | 4 | 386 | ||
NM_001320948.1 | NP_001307877.1 | 4 | 386 | |||
XM_011531433.1 | XP_011529735.1 | 4 | 346 | |||
XM_017030056.1 | XP_016885545.1 | 4 | 346 | |||
XM_017030057.1 | XP_016885546.1 | 3 | 162 | |||
GenBank mRNA | BC136862.1 | - | 4 | 299 | 9020853 | |
BC140758.1 | - | 4 | 299 | 9021626 | ||
BC144459.1 | - | 4 | 299 | 9052985 |
Gene Symbol: RBMY1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006120.3 | NP_001006120.2 | 4 | 386 | ||
NM_001320949.1 | NP_001307878.1 | 4 | 386 | |||
NM_001320950.1 | NP_001307879.1 | 5 | 437 | |||
XM_011531486.2 | XP_011529788.1 | 4 | 259 | |||
XM_011531487.2 | XP_011529789.1 | 4 | 346 | |||
XM_011531488.2 | XP_011529790.1 | 4 | 346 | |||
XM_011531489.1 | XP_011529791.1 | 4 | 346 | |||
GenBank mRNA | BC144422.1 | - | 4 | 299 | 9052943 | |
BC144423.1 | - | 5 | 350 |
Gene Symbol: RBMY1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006118.2 | NP_001006118.2 | 4 | 346 | ||
XM_011531493.1 | XP_011529795.1 | 4 | 346 | |||
XM_011531494.2 | XP_011529796.1 | 4 | 346 | |||
XM_011531495.1 | XP_011529797.1 | 4 | 346 | |||
XM_017030058.1 | XP_016885547.1 | 3 | 162 |
Gene Symbol: RBMY1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006117.3 | NP_001006117.2 | 4 | 411 | ||
XM_011531500.1 | XP_011529802.1 | 4 | 282 | |||
XM_011531502.1 | XP_011529804.1 | 4 | 282 | |||
XM_017030059.1 | XP_016885548.1 | 3 | 162 | |||
GenBank mRNA | AK313597.1 | - | 4 | 415 | ||
BC144577.1 | - | 4 | 299 | 9053106 | ||
BC152392.1 | - | 3 | 256 | 8860211 |
Gene Symbol: RBMY2FP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002193.2 | 4 | 350 | |||
GenBank mRNA | BC026077.2 | - | 4 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
