siRNA Details
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 4 | 429 | ||
GenBank mRNA | AK130965.1 | - | 4 | 418 | ||
AK312123.1 | - | 4 | 418 | |||
BC014459.1 | - | 1 | 409 | 4872948 | ||
BC058041.1 | - | 4 | 421 | 6716166 | ||
BG476195.1 | - | 4 | 538 | |||
BP382007.1 | - | 4 | 429 | |||
L13799.1 | - | 3 | 379 | |||
U02032.1 | - | 4 | 407 | |||
U37230.1 | - | 1 | 428 | |||
U43701.1 | - | 4 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
