siRNA Details
Gene Symbol: RPL23AP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC146939.1 | - | 1 | 560 | 9056867 | |
BC146942.1 | - | 1 | 560 | 9056870 |
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 4, 5 | 484 | ||
GenBank mRNA | AK130965.1 | - | 4, 5 | 473 | ||
AK312123.1 | - | 4, 5 | 473 | |||
BC014459.1 | - | 1 | 464 | 4872948 | ||
BC058041.1 | - | 4, 5 | 476 | 6716166 | ||
BG476195.1 | - | 4, 5 | 593 | |||
BP382007.1 | - | 4, 5 | 484 | |||
L13799.1 | - | 3, 4 | 434 | |||
U02032.1 | - | 4, 5 | 462 | |||
U37230.1 | - | 1 | 483 | |||
U43701.1 | - | 4, 5 | 481 |
Gene Symbol: RPL23AP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003572.2 | 4 | 878 | |||
GenBank mRNA | AK127000.1 | - | 3 | 605 | ||
BC052779.1 | - | 5 | 936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
