siRNA Details
Gene Symbol: RPL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000984.5 | NP_000975.2 | 1, 2 | 43 | ||
GenBank mRNA | AK130965.1 | - | 1, 2 | 32 | ||
AK312123.1 | - | 1, 2 | 32 | |||
BC014459.1 | - | 1 | 23 | 4872948 | ||
BC058041.1 | - | 1, 2 | 35 | 6716166 | ||
BE731473.1 | - | 1, 2 | 611 | |||
BG476195.1 | - | 1, 2 | 152 | |||
BP382007.1 | - | 1, 2 | 43 | |||
U02032.1 | - | 1, 2 | 21 | |||
U37230.1 | - | 1 | 42 | |||
U43701.1 | - | 1, 2 | 40 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
