siRNA Details
Gene Symbol: CFHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002113.2 | NP_002104.2 | 6 | 1154 | ||
XM_011509457.2 | XP_011507759.1 | 5 | 960 | |||
GenBank mRNA | AK290830.1 | - | 6 | 1144 | ||
BC016755.1 | - | 6 | 1132 | 3934474 | ||
BC107771.1 | - | 6 | 1150 | 4712577 | ||
M65292.1 | - | 6 | 1117 | |||
M65293.1 | - | 6 | 1115 | |||
X56209.1 | - | 6 | 1032 | |||
X56210.1 | - | 6 | 1080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
