siRNA Details
Gene Symbol: CFHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002113.2 | NP_002104.2 | 4 | 604 | ||
XM_011509457.2 | XP_011507759.1 | 3 | 410 | |||
GenBank mRNA | AK290830.1 | - | 4 | 594 | ||
BC016755.1 | - | 4 | 582 | 3934474 | ||
BC107771.1 | - | 4 | 600 | 4712577 | ||
BG568702.1 | - | 4 | 601 | |||
M65292.1 | - | 4 | 567 | |||
M65293.1 | - | 4 | 565 | |||
X56209.1 | - | 4 | 482 | |||
X56210.1 | - | 4 | 530 |
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000186.3 | NP_000177.2 | 20 | 3433 | ||
GenBank mRNA | AK226113.1 | - | 14 | 6301 | ||
AK291395.1 | - | 20 | 3272 | |||
BC142699.1 | - | 20 | 3245 | 40148771 | ||
M17517.1 | - | 9 | 1472 | |||
M65294.1 | - | 1 | 55 | |||
Y00716.1 | - | 20 | 3266 |
Gene Symbol: CFHR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BG618529.1 | - | 3 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
