siRNA Details
Gene Symbol: CFHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002113.2 | NP_002104.2 | 6 | 935 | ||
XM_011509457.2 | XP_011507759.1 | 5 | 741 | |||
GenBank mRNA | AK290830.1 | - | 6 | 925 | ||
BC016755.1 | - | 6 | 913 | 3934474 | ||
BC107771.1 | - | 6 | 931 | 4712577 | ||
M65292.1 | - | 6 | 898 | |||
M65293.1 | - | 6 | 896 | |||
X56209.1 | - | 6 | 813 | |||
X56210.1 | - | 6 | 861 |
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000186.3 | NP_000177.2 | 22 | 3764 | ||
GenBank mRNA | AK226113.1 | - | 16 | 6632 | ||
AK291395.1 | - | 22 | 3603 | |||
BC142699.1 | - | 22 | 3576 | 40148771 | ||
M17517.1 | - | 11 | 1803 | |||
M65294.1 | - | 3 | 386 | |||
Y00716.1 | - | 22 | 3597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
