siRNA Details
Gene Symbol: HBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000517.4 | NP_000508.1 | 1 | 124 |
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000558.4 | NP_000549.1 | 1 | 124 | ||
GenBank mRNA | AF097635.1 | - | 1 | 81 | ||
AF105974.1 | - | 1 | 79 | |||
AF147332.1 | - | 1 | 76 | |||
AF349571.1 | - | 1 | 83 | |||
AK223392.1 | - | 1 | 101 | |||
BC005931.1 | - | 1 | 95 | 4047053 | ||
BC008572.1 | - | 1 | 81 | 4177243 | ||
BC032122.2 | - | 1 | 81 | 4849981 | ||
BC050661.1 | - | 1 | 81 | 5209215 | ||
BC101846.1 | - | 1 | 88 | 8069352 | ||
BC101848.1 | - | 1 | 88 | 8069354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
