siRNA Details
Gene Symbol: ARL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113738.1 | NP_001107210.1 | 4 | 528 | ||
XM_011524875.2 | XP_011523177.1 | 4 | 581 | |||
XM_011524876.2 | XP_011523178.1 | 4 | 580 | |||
XM_011524877.2 | XP_011523179.1 | 4 | 576 | |||
XM_017024729.1 | XP_016880218.1 | 4 | 580 | |||
GenBank mRNA | AB209344.1 | - | 4 | 459 | ||
AK289590.1 | - | 4 | 466 | |||
AK315857.1 | - | 4 | 467 | |||
BC000924.3 | - | 1 | 131 | |||
BC030570.1 | - | 4 | 499 | 5518676 | ||
BC041803.1 | - | 4 | 485 | 5303659 | ||
BC071681.1 | - | 2 | 193 | |||
BX647266.1 | - | 4 | 488 |
Gene Symbol: ARL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039083.3 | NP_001034172.3 | 4 | 499 | ||
XM_011524160.2 | XP_011522462.1 | 4 | 599 | |||
XM_011524161.2 | XP_011522463.1 | 4 | 600 | |||
XM_011524162.2 | XP_011522464.1 | 4 | 600 | |||
XM_011524163.2 | XP_011522465.1 | 4 | 600 | |||
XM_011524164.2 | XP_011522466.1 | 4 | 596 | |||
XM_011524165.2 | XP_011522467.1 | 4 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
