siRNA Details
Gene Symbol: NDFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030571.3 | NP_085048.1 | 8 | 2407 | ||
GenBank mRNA | AK075495.1 | - | 5 | 378 | ||
AK124884.1 | - | 2 | 3211 | |||
AL832993.1 | - | 5 | 1888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
