siRNA Details
Gene Symbol: NDFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030571.3 | NP_085048.1 | 3 | 386 | ||
GenBank mRNA | AB097010.1 | - | 3 | 322 | ||
AB097037.1 | - | 3 | 322 | |||
AI929639.1 | - | 3 | 386 | |||
AK075495.1 | - | 4 | 306 | |||
AK075524.1 | - | 3 | 325 | |||
AK315481.1 | - | 3 | 331 | |||
AY192728.1 | - | 3 | 342 | |||
BC004317.1 | - | 3 | 274 | 3628100 | ||
DA236494.1 | - | 3 | 386 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
