siRNA Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318485.1 | NP_001305414.1 | 4 | 955 | ||
NM_017971.3 | NP_060441.2 | 4 | 610 | |||
GenBank mRNA | AB049644.1 | - | 4 | 541 | ||
AK000886.1 | - | 3 | 1566 | |||
BC009515.1 | - | 4 | 578 | 3542715 | ||
BC014316.1 | - | 4 | 547 | 4045076 | ||
BC059945.1 | - | 4 | 528 | 5397911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
