siRNA Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017971.3 | NP_060441.2 | 3, 4 | 364 | ||
GenBank mRNA | AB049644.1 | - | 3, 4 | 295 | ||
AK000886.1 | - | 2, 3 | 1320 | |||
AK315794.1 | - | 3, 4 | 298 | |||
BC009515.1 | - | 3, 4 | 332 | 3542715 | ||
BC014316.1 | - | 3, 4 | 301 | 4045076 | ||
BC059945.1 | - | 3, 4 | 282 | 5397911 | ||
BG700526.1 | - | 3, 4 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
