siRNA Details
Gene Symbol: KRTAP9-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033191.2 | NP_149461.2 | 1 | 394 | ||
GenBank mRNA | AJ406948.1 | - | 1 | 394 | ||
BC121094.2 | - | 1 | 387 | 40120009 |
Gene Symbol: KRTAP9-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031961.2 | NP_114167.2 | 1 | 412 | ||
GenBank mRNA | AJ406946.1 | - | 1 | 412 | ||
BC069560.1 | - | 1 | 404 | |||
BC110619.2 | - | 1 | 421 | 40035092 | ||
BC117287.1 | - | 1 | 433 | 40125838 |
Gene Symbol: KRTAP9-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031962.2 | NP_114168.1 | 1 | 399 | ||
GenBank mRNA | AJ406947.1 | - | 1 | 399 | ||
BC074918.2 | - | 1 | 391 | 30915341 | ||
BC074919.2 | - | 1 | 391 | 30915560 |
Gene Symbol: KRTAP9-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277332.1 | NP_001264261.1 | 1 | 390 |
Gene Symbol: KRTAP9-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031963.2 | NP_114169.2 | 1 | 394 | ||
GenBank mRNA | AJ406950.1 | - | 1 | 394 | ||
BC074981.2 | - | 1 | 385 | 30915515 | ||
BC074982.2 | - | 1 | 385 | 30915299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
