siRNA Details
Gene Symbol: KRTAP9-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031963.2 | NP_114169.2 | 1 | 190 | ||
GenBank mRNA | AJ406950.1 | - | 1 | 190 | ||
BC074981.2 | - | 1 | 181 | 30915515 | ||
BC074982.2 | - | 1 | 181 | 30915299 |
Gene Symbol: KRTAP9-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190460.1 | NP_001177389.1 | 1 | 171 | ||
XM_005257633.1 | XP_005257690.1 | 1 | 171 |
Gene Symbol: KRTAP9-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031961.2 | NP_114167.2 | 1 | 178 | ||
GenBank mRNA | BC069560.1 | - | 1 | 170 | ||
BC117287.1 | - | 1 | 199 | 40125838 |
Gene Symbol: KRTAP9-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031962.2 | NP_114168.1 | 1 | 195 | ||
GenBank mRNA | AJ406947.1 | - | 1 | 195 | ||
BC074918.2 | - | 1 | 187 | 30915341 | ||
BC074919.2 | - | 1 | 187 | 30915560 |
Gene Symbol: KRTAP9-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033191.2 | NP_149461.2 | 1 | 205 | ||
GenBank mRNA | AJ406948.1 | - | 1 | 205 | ||
BC121094.2 | - | 1 | 198 | 40120009 |
Gene Symbol: KRTAP9-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277331.1 | NP_001264260.1 | 1 | 171 |
Gene Symbol: KRTAP9-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277332.1 | NP_001264261.1 | 1 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
