siRNA Details
Gene Symbol: CHD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256336.1 | NP_001243265.1 | 6 | 597 | ||
NM_004284.4 | NP_004275.4 | 7 | 704 | |||
NM_024568.2 | NP_078844.2 | 7 | 676 | |||
NR_046070.1 | 7 | 704 | ||||
XM_006711639.3 | XP_006711702.1 | 6 | 572 | |||
XM_017002858.1 | XP_016858347.1 | 8 | 1432 | |||
XM_017002859.1 | XP_016858348.1 | 7 | 559 | |||
XM_017002860.1 | XP_016858349.1 | 7 | 645 | |||
XM_017002861.1 | XP_016858350.1 | 7 | 652 | |||
XM_017002863.1 | XP_016858352.1 | 6 | 467 | |||
XM_017002864.1 | XP_016858353.1 | 7 | 645 | |||
GenBank mRNA | AF054177.1 | - | 7 | 625 | ||
AF537213.1 | - | 7 | 661 | |||
AK001342.1 | - | 7 | 661 | |||
AK027631.1 | - | 7 | 633 | |||
AK223496.1 | - | 5 | 355 | |||
AK298424.1 | - | 6 | 537 | |||
AK307400.1 | - | 7 | 661 | |||
BC001171.1 | - | 7 | 638 | |||
BC005038.1 | - | 7 | 601 | |||
BC043501.1 | - | 7 | 622 | |||
CN255649.1 | - | 4 | 277 | |||
EF560738.1 | - | 7 | 672 | |||
KJ534809.1 | - | 7 | 641 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
