siRNA Details
Gene Symbol: CHD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256336.1 | NP_001243265.1 | 5 | 486 | ||
NM_001256337.1 | NP_001243266.1 | 2 | 226 | |||
NM_004284.4 | NP_004275.4 | 6 | 593 | |||
NM_024568.2 | NP_078844.2 | 6 | 565 | |||
NR_046070.1 | 6 | 593 | ||||
XM_006711639.3 | XP_006711702.1 | 5 | 461 | |||
XM_017002858.1 | XP_016858347.1 | 7 | 1321 | |||
XM_017002859.1 | XP_016858348.1 | 6 | 448 | |||
XM_017002860.1 | XP_016858349.1 | 6 | 541 | |||
XM_017002861.1 | XP_016858350.1 | 6 | 541 | |||
XM_017002863.1 | XP_016858352.1 | 5 | 356 | |||
XM_017002864.1 | XP_016858353.1 | 6 | 541 | |||
GenBank mRNA | AF054177.1 | - | 6 | 514 | ||
AF537213.1 | - | 6 | 550 | |||
AK001342.1 | - | 6 | 550 | |||
AK027631.1 | - | 6 | 522 | |||
AK223496.1 | - | 4 | 244 | |||
AK293157.1 | - | 2 | 161 | |||
AK298424.1 | - | 5 | 426 | |||
AK307400.1 | - | 6 | 550 | |||
BC001171.1 | - | 6 | 527 | |||
BC005038.1 | - | 6 | 490 | |||
BC043501.1 | - | 6 | 511 | |||
CN255649.1 | - | 3 | 166 | |||
DA745071.1 | - | 6 | 544 | |||
EF560738.1 | - | 6 | 552 | |||
KJ534809.1 | - | 6 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
