siRNA Details
Gene Symbol: ANKRD20A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012419.2 | NP_001012419.1 | 4 | 646 | ||
XM_005272525.3 | XP_005272582.1 | 4 | 902 | |||
XM_011543860.2 | XP_011542162.1 | 4 | 903 | |||
XM_011543861.2 | XP_011542163.1 | 4 | 907 | |||
GenBank mRNA | BC140809.1 | - | 4 | 646 | 9021677 | |
BC171793.1 | - | 4 | 646 | 9054447 |
Gene Symbol: ANKRD20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012421.1 | NP_001012421.1 | 4 | 680 | ||
XM_011517877.2 | XP_011516179.1 | 4 | 680 | |||
XM_011517878.2 | XP_011516180.1 | 4 | 680 | |||
XM_011517879.2 | XP_011516181.1 | 4 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
