siRNA Details
Gene Symbol: HMGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313892.1 | NP_001300821.1 | 5 | 735 | ||
NM_001313893.1 | NP_001300822.1 | 5 | 1495 | |||
NM_002128.5 | NP_002119.1 | 5 | 738 | |||
GenBank mRNA | AB208998.1 | - | 4 | 1627 | ||
AK057120.1 | - | 5 | 601 | |||
AK122825.1 | - | 5 | 1497 | |||
AK291494.1 | - | 5 | 709 | |||
AK310975.1 | - | 5 | 556 | |||
AX795278.1 | - | 5 | 630 | |||
AY377859.1 | - | 4 | 554 | |||
BC003378.1 | - | 5 | 630 | 2901382 | ||
BC030981.1 | - | 5 | 649 | 4041682 | ||
BC066889.1 | - | 5 | 633 | 5268698 | ||
BC067732.1 | - | 5 | 653 | 4818524 | ||
BC141844.1 | - | 5 | 631 | 8860319 | ||
BT006940.1 | - | 4 | 554 | |||
BT020159.1 | - | 4 | 554 | |||
BX647267.1 | - | 5 | 1168 | |||
CR456863.1 | - | 4 | 554 | |||
CR749614.1 | - | 5 | 726 | |||
D63874.1 | - | 5 | 630 | |||
X12597.1 | - | 5 | 606 |
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137482.1 | - | 1 | 589 | 9021482 | |
BC137483.1 | - | 1 | 589 | 9021483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
