siRNA Details
Gene Symbol: GNG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005274.2 | NP_005265.1 | 3 | 511 | ||
GenBank mRNA | AF038955.1 | - | 2 | 194 | ||
AF493873.1 | - | 2 | 157 | |||
AK312111.1 | - | 2 | 270 | |||
BC003563.1 | - | 2 | 240 | 3502879 | ||
BM809260.1 | - | 2 | 250 | |||
BT006823.1 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
