siRNA Details
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024448.2 | 11 | 2905 | |||
GenBank mRNA | AK056875.1 | - | 11 | 2835 | ||
AK091837.1 | - | 10 | 2643 | |||
AK125808.1 | - | 4 | 3305 | |||
BC063045.1 | - | 9 | 1148 | |||
CR933599.1 | - | 3 | 3177 | |||
S82637.1 | - | 3 | 397 | |||
X52204.1 | - | 3 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
