siRNA Details
Gene Symbol: SAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127380.2 | NP_001120852.1 | 3 | 235 | ||
NM_030754.4 | NP_110381.2 | 3 | 235 | |||
GenBank mRNA | AK307163.1 | - | 3 | 195 | ||
BC020795.1 | - | 3 | 196 | 4767650 | ||
M23699.1 | - | 2 | 159 | |||
M23700.1 | - | 2 | 159 | |||
M26152.1 | - | 3 | 182 | |||
X51445.1 | - | 2 | 150 |
Gene Symbol: SAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000331.5 | NP_000322.2 | 3 | 382 | ||
NM_001178006.2 | NP_001171477.1 | 4 | 296 | |||
NM_199161.4 | NP_954630.1 | 3 | 235 | |||
GenBank mRNA | BC007022.1 | - | 3 | 196 | 4071612 | |
BC105796.1 | - | 4 | 261 | 4722684 | ||
BG533276.1 | - | 344 | ||||
BG562565.1 | - | 3 | 235 | |||
BG565078.1 | - | 3 | 344 | |||
BG567902.1 | - | 287 | ||||
BQ691948.1 | - | 2 | 98 | |||
BX102362.1 | - | 3 | 302 | |||
CD102084.1 | - | 3 | 199 | |||
CR542241.1 | - | 2 | 159 | |||
M10906.1 | - | 2 | 159 | |||
M23698.1 | - | 2 | 159 | |||
X51443.1 | - | 2 | 138 |
Gene Symbol: SAA2-SAA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199744.1 | NP_001186673.1 | 3 | 235 | ||
GenBank mRNA | BG616641.1 | - | 3 | 197 | ||
BX117371.1 | - | 3 | 179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
