siRNA Details
Gene Symbol: OCM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097622.1 | NP_001091091.1 | 2, 3 | 280 | ||
XM_011515485.2 | XP_011513787.1 | 3, 4 | 227 | |||
GenBank mRNA | BC069468.1 | - | 2, 3 | 188 | 7262729 | |
BC069512.1 | - | 2, 3 | 188 | 7262741 | ||
BC069550.1 | - | 2, 3 | 188 | 7262753 | ||
BC113704.1 | - | 2, 3 | 280 | 8322756 | ||
BC113706.1 | - | 2, 3 | 280 | 8322758 | ||
BC147028.1 | - | 2, 3 | 282 | 9056956 | ||
BC147029.1 | - | 2, 3 | 282 | 9056957 |
Gene Symbol: OCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006188.3 | NP_006179.2 | 2, 3 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
