siRNA Details
Gene Symbol: KHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251874.1 | NP_001238803.1 | 4 | 965 | ||
NM_030568.4 | NP_085045.3 | 3 | 681 | |||
GenBank mRNA | BC022080.2 | - | 3 | 651 | 4813061 | |
BM725230.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
