siRNA Details
Gene Symbol: CBWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172003.3 | NP_742000.1 | 15 | 1583 | ||
XM_005263600.2 | XP_005263657.1 | 14 | 1526 | |||
XM_005263602.2 | XP_005263659.1 | 14 | 1496 | |||
XM_005263603.2 | XP_005263660.1 | 16 | 1617 | |||
XM_005263604.2 | XP_005263661.1 | 13 | 1439 | |||
XM_011510667.1 | XP_011508969.1 | 14 | 1453 | |||
XM_011510668.2 | XP_011508970.1 | 12 | 1143 | |||
XM_011510669.2 | XP_011508971.1 | 16 | 1627 | |||
XM_017003408.1 | XP_016858897.1 | 17 | 1747 | |||
GenBank mRNA | AF452722.1 | - | 15 | 1439 | ||
AK124867.1 | - | 6 | 3148 | |||
AY343910.1 | - | 15 | 1468 | |||
BC120991.1 | - | 15 | 1454 | 40116409 | ||
BC120992.1 | - | 15 | 1454 | 40116411 | ||
DA296279.1 | - | 1 | 322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
