siRNA Details
Gene Symbol: CFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270420.1 | NP_001257349.1 | 5 | 901 | ||
NM_001270421.1 | NP_001257350.1 | 4 | 791 | |||
NM_032545.3 | NP_115934.1 | 6 | 1016 | |||
GenBank mRNA | AF312769.1 | - | 6 | 774 | ||
BC110080.1 | - | 6 | 889 | 40030641 | ||
BG654700.1 | - | 3 | 472 | |||
CA948246.1 | - | 1 | 126 |
Gene Symbol: CFC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079530.1 | NP_001072998.1 | 6 | 1119 | ||
GenBank mRNA | BM716751.1 | - | 3 | 391 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
