siRNA Details
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY894991.1 | - | 2 | 274 | ||
BC062732.1 | - | 2 | 248 | |||
BC070334.1 | - | 2 | 305 | |||
BC070336.1 | - | 2 | 309 | |||
BC073763.1 | - | 2 | 294 | |||
BC073764.1 | - | 2 | 291 |
Gene Symbol: IGKV1D-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | U86790.1 | - | 2 | 280 |
Gene Symbol: IGKV1D-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035035.1 | - | 1 | 214 |
Gene Symbol: IGKV@ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF103379.1 | - | 1 | 202 | ||
AF103383.1 | - | 2 | 203 |
Gene Symbol: SCFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13056.1 | - | 610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
