siRNA Details
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018.4 | NP_001009.1 | 2 | 77 | ||
NM_001308226.1 | NP_001295155.1 | 2 | 168 | |||
GenBank mRNA | BC064908.1 | - | 1 | 21 | 6053159 | |
BC105810.1 | - | 2 | 55 | 6744553 | ||
BC141832.1 | - | 1 | 21 | 40148744 | ||
BG253776.1 | - | 1 | 16 | |||
BG705752.1 | - | 2 | 175 | |||
BM558707.1 | - | 2 | 156 | |||
BU602036.1 | - | 2 | 78 | |||
J02984.1 | - | 2 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
