siRNA Details
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018.4 | NP_001009.1 | 4 | 412 | ||
NM_001308226.1 | NP_001295155.1 | 4 | 503 | |||
GenBank mRNA | AF145025.1 | - | 1 | 372 | ||
BC064908.1 | - | 1 | 356 | 6053159 | ||
BC105810.1 | - | 4 | 390 | 6744553 | ||
BC141832.1 | - | 1 | 356 | 40148744 | ||
BE798984.1 | - | 4 | 591 | |||
BG253776.1 | - | 1 | 351 | |||
BG705752.1 | - | 4 | 509 | |||
BM558707.1 | - | 4 | 491 | |||
BU602036.1 | - | 4 | 413 | |||
J02984.1 | - | 4 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
