siRNA Details
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 5 | 524 | ||
NM_173614.2 | NP_775885.1 | 5 | 524 | |||
XM_017023156.1 | XP_016878645.1 | 5 | 571 | |||
GenBank mRNA | AK225803.1 | - | 5 | 494 | ||
AL832855.1 | - | 5 | 518 | |||
BC041131.1 | - | 5 | 498 | 5757234 | ||
BG830626.1 | - | 530 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 5 | 518 | ||
GenBank mRNA | BC065535.1 | - | 5 | 471 | 6007603 | |
X57398.1 | - | 5 | 356 |
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 5 | 587 | ||
XM_005255318.1 | XP_005255375.1 | 5 | 581 | |||
GenBank mRNA | BI459221.1 | - | 5 | 561 | ||
BI596373.1 | - | 5 | 516 | |||
BU169681.1 | - | 470 | ||||
BU526741.1 | - | 5 | 524 | |||
BX383108.2 | - | 5 | 508 | |||
DR760717.1 | - | 5 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
