siRNA Details
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 7 | 775 | ||
GenBank mRNA | BC065535.1 | - | 7 | 728 | 6007603 | |
X57398.1 | - | 7 | 613 |
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 7 | 781 | ||
NM_173614.2 | NP_775885.1 | 7 | 781 | |||
XM_017023156.1 | XP_016878645.1 | 7 | 828 | |||
GenBank mRNA | AK225803.1 | - | 7 | 751 | ||
AL832855.1 | - | 7 | 775 | |||
BC028389.1 | - | 6 | 641 | 4824724 | ||
BC041131.1 | - | 7 | 755 | 5757234 |
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 7 | 844 | ||
XM_005255318.1 | XP_005255375.1 | 7 | 838 | |||
GenBank mRNA | BQ954152.1 | - | 46 | |||
BX383108.2 | - | 7 | 765 | |||
CD653707.1 | - | 1 | 130 | |||
DA392676.1 | - | 2 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
