siRNA Details
Gene Symbol: NOMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004067.3 | NP_001004067.1 | 7, 8 | 867 | ||
XM_005255318.1 | XP_005255375.1 | 7, 8 | 861 | |||
GenBank mRNA | BQ954152.1 | - | 69 | |||
BX383108.2 | - | 7, 8 | 788 | |||
CD653707.1 | - | 1, 2 | 153 | |||
DA392676.1 | - | 2, 3 | 196 |
Gene Symbol: NOMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014287.3 | NP_055102.3 | 7, 8 | 798 | ||
GenBank mRNA | BC065535.1 | - | 7, 8 | 751 | 6007603 | |
X57398.1 | - | 7, 8 | 636 |
Gene Symbol: NOMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004060.1 | NP_001004060.1 | 7, 8 | 804 | ||
NM_173614.2 | NP_775885.1 | 7, 8 | 804 | |||
XM_017023156.1 | XP_016878645.1 | 7, 8 | 851 | |||
GenBank mRNA | AK225803.1 | - | 7, 8 | 774 | ||
AL832855.1 | - | 7, 8 | 798 | |||
BC028389.1 | - | 6, 7 | 664 | 4824724 | ||
BC041131.1 | - | 7, 8 | 778 | 5757234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
