siRNA Details
Gene Symbol: FTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000146.3 | NP_000137.2 | 4 | 711 | ||
GenBank mRNA | AF147331.1 | - | 3 | 322 | ||
AK026534.1 | - | 1 | 711 | |||
AK130191.1 | - | 4 | 711 | |||
AK130205.1 | - | 4 | 706 | |||
AK131048.1 | - | 4 | 711 | |||
AK131050.1 | - | 4 | 711 | |||
AK131053.1 | - | 4 | 725 | |||
AK307065.1 | - | 4 | 701 | |||
AK311773.1 | - | 4 | 711 | |||
AY207005.1 | - | 4 | 715 | |||
AY466472.1 | - | 4 | 514 | |||
BC002991.1 | - | 4 | 679 | 2905327 | ||
BC004245.1 | - | 4 | 700 | 3615360 | ||
BC008439.1 | - | 1 | 711 | 4093482 | ||
BC008441.1 | - | 1 | 711 | |||
BC013928.1 | - | 1 | 715 | 3839120 | ||
BC016346.1 | - | 1 | 715 | 4066010 | ||
BC016354.1 | - | 4 | 715 | 4092360 | ||
BC016715.1 | - | 1 | 715 | 4067055 | ||
BC018990.2 | - | 1 | 683 | 3503710 | ||
BC021670.1 | - | 1 | 714 | 4051089 | ||
BC058820.1 | - | 1 | 704 | 4049525 | ||
BC062708.1 | - | 1 | 714 | 4774403 | ||
BX571748.1 | - | 1 | 710 | |||
CR456715.1 | - | 4 | 512 | |||
M10119.1 | - | 4 | 591 | |||
M11147.1 | - | 4 | 663 | |||
M12938.1 | - | 3 | 416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
