siRNA Details
Gene Symbol: CROCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014675.4 | NP_055490.4 | 16 | 2327 | ||
XM_006711058.3 | XP_006711121.1 | 15 | 2118 | |||
XM_011542468.1 | XP_011540770.1 | 16 | 2304 | |||
XM_011542470.2 | XP_011540772.1 | 16 | 2304 | |||
XM_017002913.1 | XP_016858402.1 | 16 | 2304 | |||
XM_017002914.1 | XP_016858403.1 | 16 | 2304 | |||
XM_017002915.1 | XP_016858404.1 | 16 | 2304 | |||
GenBank mRNA | AB007914.2 | - | 14 | 1966 | ||
BC126911.1 | - | 13 | 1848 | |||
BC126912.1 | - | 13 | 1848 | |||
DQ139275.1 | - | 3 | 1684 |
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026752.1 | 4 | 467 | |||
GenBank mRNA | AK090414.1 | - | 9 | 1248 | ||
BC006312.2 | - | 4 | 514 | |||
BC047736.1 | - | 4 | 467 |
Gene Symbol: CROCCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023386.1 | 13 | 2413 | |||
GenBank mRNA | AB067509.1 | - | 13 | 2412 | ||
BC033082.1 | - | 10 | 1346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
