siRNA Details
Gene Symbol: CROCCP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023386.1 | 14 | 2489 | |||
GenBank mRNA | AB067509.1 | - | 14 | 2488 | ||
BC033082.1 | - | 11 | 1422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
