siRNA Details
Gene Symbol: ZNF33B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305033.1 | NP_001291962.1 | 3 | 531 | ||
NM_001305035.1 | NP_001291964.1 | 4 | 584 | |||
NM_001305036.1 | NP_001291965.1 | 4 | 589 | |||
NM_001305037.1 | NP_001291966.1 | 3 | 439 | |||
NM_001305038.1 | NP_001291967.1 | 3 | 531 | |||
NM_001305039.1 | NP_001291968.1 | 4 | 589 | |||
NM_001305040.1 | NP_001291969.1 | 4 | 584 | |||
NM_006955.2 | NP_008886.1 | 4 | 584 | |||
NR_130948.1 | 4 | 584 | ||||
NR_130949.1 | 3 | 439 | ||||
NR_130950.1 | 3 | 439 | ||||
NR_130951.1 | 2 | 386 | ||||
GenBank mRNA | AJ491697.1 | - | 4 | 291 | ||
AK096683.1 | - | 3 | 155 | |||
AK309564.1 | - | 4 | 316 | |||
AK310073.1 | - | 4 | 584 | |||
BC048313.2 | - | 4 | 289 | |||
BC065181.1 | - | 4 | 266 | |||
BC107726.1 | - | 4 | 282 | |||
BG536481.1 | - | 3 | 248 | |||
CD652356.1 | - | 3 | 258 | |||
DA230792.1 | - | 4 | 321 | |||
DA493121.1 | - | 4 | 293 | |||
DA826003.1 | - | 2 | 113 | |||
DA959465.1 | - | 3 | 171 | |||
DB156590.1 | - | 3 | 171 | |||
DR000119.1 | - | 4 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
