siRNA Details
Gene Symbol: OVOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020287.1 | XP_016875776.1 | 29 | 3906 | ||
GenBank mRNA | BX647938.1 | - | 10 | 6354 |
Gene Symbol: LOC728715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020286.1 | XP_016875775.1 | 4 | 456 |
Gene Symbol: OVOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020288.1 | XP_016875777.1 | 30 | 4086 | ||
GenBank mRNA | AL831947.1 | - | 2137 | |||
BC039117.1 | - | 4 | 1012 | |||
BC046190.1 | - | 3 | 361 | |||
CR749395.1 | - | 3 | 2325 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
