siRNA Details
Gene Symbol: ZNF702P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003578.1 | 4 | 927 | |||
GenBank mRNA | AK023047.1 | - | 1 | 140 | ||
AK092593.1 | - | 4 | 927 | |||
AL832505.1 | - | 4 | 924 | |||
BC032590.1 | - | 1 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
