siRNA Details
Gene Symbol: ZNF845 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321522.1 | NP_001308451.1 | 5 | 2432 | ||
NM_001321523.1 | NP_001308452.1 | 5 | 2371 | |||
NM_001321524.1 | NP_001308453.1 | 5 | 2379 | |||
NM_138374.2 | NP_612383.1 | 4 | 2277 | |||
GenBank mRNA | AK096653.1 | - | 1 | 700 | ||
BC007307.1 | - | 1 | 1133 |
Gene Symbol: ZNF702P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003578.1 | 4 | 1109 | |||
GenBank mRNA | AK023047.1 | - | 1 | 322 | ||
AK092593.1 | - | 4 | 1109 | |||
AL832505.1 | - | 4 | 1106 | |||
BC032590.1 | - | 1 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
