siRNA Details
Gene Symbol: CHCHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320327.1 | NP_001307256.1 | 3 | 581 | ||
NM_016139.3 | NP_057223.1 | 3 | 581 | |||
GenBank mRNA | AF078845.1 | - | 3 | 445 | ||
AY605046.1 | - | 3 | 364 | |||
AY633613.1 | - | 3 | 385 | |||
BC003079.2 | - | 3 | 401 | 2988156 | ||
BC015639.2 | - | 3 | 385 | 4637353 | ||
BC066331.1 | - | 3 | 404 | 4792941 | ||
BC071985.1 | - | 3 | 401 | 6426679 | ||
BC100275.1 | - | 3 | 397 | 6192342 | ||
BF347122.1 | - | 456 | ||||
CN363208.1 | - | 3 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
